Zinc chloride ≥97.0%, granulated USP
Supplier: SPECTRUM CHEMICAL MFG CORP
|
|
Synonyms:
Zinc dichloride, Butter of zinc, Dichlorozinc, Zinc (II) chloride
ZI105-100GM
ZI105-2.5KG
ZI105-500GM
ZI105-12KG
ZI105-1.5KGBPL
ZI105-50KGBL
95031-392EA
245.86
USD
95031-392
95031-394
95031-396
89079-050
75812-640
75812-642
Zinc chloride ≥97.0%, granulated USP
Zinc chloride
Zinc Chloride, Granular, USP - All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
GHS hazard : Corrosion;Exclamation mark
UN2331
Formula:
ZnCl₂ Storage Temperature: Ambient |
MDL Number:
MFCD00011295 CAS Number: 7646-85-7 UN: 2331 ADR: 8,III |
Specification Test Results
Assay | 97.0 - 100.5 % |
Oxychloride | To Pass Test |
Sulfates (SO₄) | max. 0.03 % |
Alkalies and Alkaline Earths | max. 1.0 % |
Ammonium Salts | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...