Grapefruit oil FCC, cold-pressed

Supplier: Spectrum Chemical Mfg. Corp.

G1047-100ML G1047-4LT G1047-500MLG G1047-500MLG
95030-752EA 111.38 USD
95030-752 95030-754 700014-338 95030-756
Grapefruit oil FCC, cold-pressed
Grapefruit oil

Grapefruit Oil, Coldpressed, FCC is an astringent oil used for both it flavoring and antimicrobial properties. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Flame;Health hazard;Exclamation mark
UN1197

Storage Temperature: Ambient
MDL Number: MFCD00240720
CAS Number: 8016-20-4
UN: 1197
ADR: 3,III

Order Now

Specification Test Results

Angular Rotation +91 to +96
Refractive Index @ 20°C 1.475 - 1.478
Residue On Evaporation 5.0 - 10.0 %
Specific Gravity 0.848 - 0.856
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR