Anti-Cytokeratin 5/14 Mouse Monoclonal Antibody [clone: XM26 + LL002]

Supplier: Diagnostic Biosystems

PDM140
95029-754EA 353.66 USD
95029-754
Anti-Cytokeratin 5/14 Mouse Monoclonal Antibody [clone: XM26 + LL002]
Antibodies
Type:
Antigen: Cytokeratin 5/14
Clonality: Monoclonal
Clone: XM26 + LL002
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1 + IgG3
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR