Ampicillin, HP Ampillicin™

Supplier: G-Biosciences

GD-6060
95029-318EA 303.54 USD
95029-318
Ampicillin, HP Ampillicin™
Ampicillin

HP Ampicillin is a stronger and more stable inhibitor of ß-lactamase than regular ampicillin and therefore supports only the growth of ampicillin resistant colonies even after prolonged incubation


  • Reduces satellite colonies
  • Lyophilised format for improved stability
  • Reconstituted 1000X concentration
  • Suitable for 5 L media or agar


Culture plates can be incubated for a longer period at 37°C, eliminating the time constraint associated with normal ampicillin.


Ordering information: HP Ampicillin is supplied lyophilised. When reconstituted in 500 µL deionised water it produces a 1000X solution. Each kit contains 10 vials, suitable for a total volume of 5 L culture media or agar.

Formula: C₁₆H₁₉N₃O₄S
MDL Number: MFCD00005175
CAS Number: 69-53-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR