EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) crosslinker

Supplier: G-Biosciences
Danger

BC25-5 BC25-1 BC25-50
95029-294EA 103.42 USD
95029-294 82021-610 76285-476
EDC-HCl (N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride) crosslinker
N-(3-Dimethylaminopropyl)-N'-ethylcarbodiimide hydrochloride

EDC is a heterobifunctional, water-soluble, zero-length carbodiimide crosslinker that is used to couple carboxyl groups to primary amines. EDC activates carboxyl groups first and forms amine reactive O-acylisourea imtermediate that spontaneously reacts with primary amines to form an amide bond and isourea by-product.


The unstable nature of the intermediate in aqueous solutions makes 2-step coupling, however in conjunction with N-hydroxysuccinimide, a 2-step coupling is possible.
EDC is ideal for peptide immobilization and hapten-carrier protein conjugation

Formula: C₈H₁₇N₃·HCl
Melting Pt: ∼ 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012503
CAS Number: 25952-53-8
EINECS: 247-361-2
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d8bf2864bcf21ffe6dcca6f22388d1fe427e22&vid=9080dd6b66a1622ef32a2da2138f85e44cc881b8&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR