Warthin-Starry Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
KT036
95027-228EA 572.08 USD
95027-228
Warthin-Starry Histology Stain Kit, Diagnostic Biosystems
Stains Histology Stains
This kit stains Helicobacter pylori, Spirochetes, Legionella pneumophila and cat scratch fever bacteria in paraffin tissue sections
Helicobacter pylori, Legionella pneumophila 
stain dark brown, while Spirochetes and cat scratch fever bacteria stain black.