Reticulum Stain, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
KT031
95027-218EA 605.21 USD
95027-218
Reticulum Stain, Diagnostic Biosystems
Stains Histology Stains
This kit stains reticulum fibers in paraffin tissue sections
The reticulum fibers stain gray to black, while the nuclei and background stain pink to rose.