P.T.A.H. Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
KT029
95027-214EA 609.41 USD
95027-214
P.T.A.H. Kit, Diagnostic Biosystems
Stains Histology Stains
This kit stains collagen, fibrin and striated muscle in paraffin tissue sections
The collagen stains brownish red to red, fibrin and muscle stain blue, while the nuclei stain dark blue.