Mucicarmine Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
KT024
95027-206EA 478.15 USD
95027-206
Mucicarmine Histology Stain Kit, Diagnostic Biosystems
Stains Histology Stains
This kit stains epithelial mucins in paraffin tissue sections
The epithelial mucin stains pink to red, while the nuclei stain blue.