Solvent Blue 38 kit stain for histology

Supplier: Diagnostic Biosystems

KT022
95027-202EA 711.67 USD
95027-202
Solvent Blue 38 kit stain for histology
Solvent Blue 38

This kit stains myelin sheath and Nissl substance in paraffin tissue sections


The myelin fibers stain blue to greenish blue, while the cells stain pink to violet.

Formula: C32H12CuN8Na2O6S2
MDL Number: MFCD00071424
CAS Number: 1328-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR