Iron Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
KT021
95027-200EA 473.51 USD
95027-200
Iron Stain Kit, Diagnostic Biosystems
Stains Histology Stains
This kit stains ferric iron in paraffin tissue sections
The iron deposits stain blue, while the nuclei and other tissue stain red and pink respectively.