Gram color staining kit
Supplier: Diagnostic Biosystems
KT018
95027-194EA 573.51 USD
95027-194
Gram color staining kit
Gram color staining kit
This kit stains Gram-positive and Gram-negative bacteria in paraffin tissue sections
The Gram-positive bacteria stain blue, Gram-negative bacteria stain red, while other tissues and the nuclei stain yellow and red respectively.