Giemsa's stain kit

Supplier: Diagnostic Biosystems

KT016
95027-192EA 561.79 USD
95027-192
Giemsa's stain kit
Giemsa's stain

This kit stains bone marrow cells, Helicobacter pylori, mast cells and certain microorganism in paraffin tissue sections


The erythrocytes stain gray, yellow or pink, H. pylori stain blue, mast cells stain dark blue with red granules, nuclei stain blue to violet and the cytoplasm stains light blue.

MDL Number: MFCD00081642
CAS Number: 51811-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR