Colloidal Iron Histology Stain Kit, Diagnostic Biosystems
Supplier: Diagnostic Biosystems
KT007
95027-178EA 474.16 USD
95027-178
Colloidal Iron Histology Stain Kit, Diagnostic Biosystems
Stains Histology Stains
The Colloidal Iron Stain Kit is designed for the histological visualization of acid mucopolysaccharides.