QSH Lab-Pal Electrophoresis System, IBI Scientific
Supplier: IBI Scientific
IB50000
95025-238EA 589.68 USD
95025-238
QSH Lab-Pal Electrophoresis System, IBI Scientific
Electrophoresis Systems Horizontal Gel Electrophoresis Systems
IBI's QSH Lab-Pal horizontal gel box is specially designed for the educational market, based upon years of tested and proven performance in the research market
The dual casting system allows casting independent from the unit, while providing maximum efficiency between lab partners. The dual gel casting system eliminates the need for casting tape. The QSH Lab-Pal casting system also allows for numerous gel slides to be cast and stored for later use.
The QSH Lab-Pal is constructed of durable cast acrylic, which allows for years of use and resists cracking or warping. The operation manual includes casting procedures, replacement parts, and helpful hints to maximize your unit's performance in your teaching lab.
The unit dimensions are 7.0Hx15.0Wx30.5Lcm and the gel dimensions are 5.0Wx7.0Lcm. The maximum sample capacity is 10 samples, the buffer capacity is 250ml, and the maximum voltage is 200V. The QSH Lab-Pal comes complete with the patented vented lid, dual casting fixture, four glass casting slides, two 1.5mm x 5 tooth combs, bubble level, power cords, and operation manual.
The QSH Lab-Pal incorporates a patented vented lid which allows heat to escape from the gel rig and thus improves the resolution of faster gel runs. Additional safety is incorporated into the lid with the patented gravity electrical connectors, which disconnects the power to the unit when the lid is removed.