Anti-HAS Mouse Monoclonal Antibody [clone: OCH1E5]

Supplier: Diagnostic Biosystems

MOB426-05 MOB426 PDM162
95024-584EA 439.74 USD
95024-584 99990-786 10064-976
Anti-HAS Mouse Monoclonal Antibody [clone: OCH1E5]
Antibodies
This antibody recognizes an uncharacterized antigen present in both adults and fetal normal hepatocytes to produce a distinct granular cytoplasmic staining. This antibody stains majority of hepatocellular carcinomas.

Type:
Antigen: Hepatocyte Specific Antigen
Clonality: Monoclonal
Clone: OCH1E5
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR