Letheen Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M414-500G
95022-910EA 251.38 USD
95022-910
Letheen Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For determining the phenol coefficient of quaternary ammonium compounds using Escherichia coli or Staphylococcus aureus ATCC 6538.
  • Microbiological culture media
  • 500 g poly bottles