Bile Salts Agar, Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M739S-500G
95022-204EA 244.94 USD
95022-204
Bile Salts Agar, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and enumeration of bile-tolerant enteric bacteria responsible for food poisoning.
  • Microbiological culture media
  • Packaged in 500 g poly bottles