Minimal Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M512-500G
95021-928EA 285.94 USD
95021-928
Minimal Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and characterization of nutritional mutants of Escherichia coli.
  • Microbiological culture media
  • 500 g poly bottles