Antibiotic Assay Medium No. 6 Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M223-500G
95021-514EA 192.86 USD
95021-514
Antibiotic Assay Medium No. 6 Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the induction of spore production in Bacillus subtilis strains used in antibiotic assay.
  • Microbiological culture media
  • Packaged in 500 g poly bottles