Chapman Stone Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M215-500G
95021-500EA 226.13 USD
95021-500
Chapman Stone Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation of Staphylococci causing food poisoning.
  • Microbiological culture media
  • 500 g poly bottles