HP6 Agar Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M1587-500G
95021-304EA 280.03 USD
95021-304
HP6 Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the isolation and cultivation of Cytophaga, Herpetosiphon, Saprospira, and Flexithrix species.
  • Microbiological culture media
  • 500 g poly bottles