Soybean Bile Broth Base, Modified, Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M1286I-500G
95020-870EA 234.71 USD
95020-870
Soybean Bile Broth Base, Modified, Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
Recommended as an enrichment medium for the detection of Escherichia coli O157:H7 from food and animal foodstuffs.
  • Microbiological culture media
  • 500 g poly bottle