FAGI Agar Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M1196-500G
95020-674EA 255.41 USD
95020-674
FAGI Agar Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the detection of Escherichia coli in water samples.
  • Microbiological culture media
  • 500 g poly bottles