MUG Tryptone Water Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M1190-500G
95020-658EA 459.79 USD
95020-658
MUG Tryptone Water Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the detection of indole producing microorganisms by fluorogenic method
  • Microbiological culture media
  • 500 g poly bottles