M-Tergitol 7 Agar Base Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M1066-500G
95020-406EA 289.62 USD
95020-406
M-Tergitol 7 Agar Base Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and identification of injured coliforms from chlorinated water using membrane filter technique.
  • Microbiological culture media
  • 500 g poly bottles