Nitrate Agar Dehydrated Culture Media, HiMedia Laboratories
Supplier: HiMedia
M072-500G
95020-184EA 178.18 USD
95020-184
Nitrate Agar Dehydrated Culture Media, HiMedia Laboratories
Media for Microbiology Dehydrated Media for Microbiology
For the detection of nitrate reduction by bacteria.
  • Microbiological culture media
  • 500 g poly bottles