MacConkey Agar without CV, with 1.2% Agar, Dehydrated Culture Media, HiMedia
Supplier: HiMedia
M008B-500G
95020-018EA 187.62 USD
95020-018
MacConkey Agar without CV, with 1.2% Agar, Dehydrated Culture Media, HiMedia
Media for Microbiology Dehydrated Media for Microbiology
For the selective isolation and differentiation of lactose fermenting and lactose nonfermenting enteric bacteria.
  • Microbiological culture media
  • 500 g poly bottles