Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))

Supplier: Cytiva
RPN4301
95019-222EA 1870.15 USD
95019-222
Anti-IgG Goat Antibody (HRP (Horseradish Peroxidase))
Antibodies
For use in ELISA systems. For ELISA, it is recommended that the conjugate should be diluted in the range 1:1000 to 1:4000.

Amdex conjugates are already supplied as components of the Biotrak high-sensitivity cytokine ELISAs, complete with fully-optimized protocols.

Type: Secondary
Antigen: IgG
Clonality:
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR