M13KO7 Helper Phage, Cytiva
Supplier: Cytiva
27-1524-01
95017-590EA 143.36 USD
95017-590
M13KO7 Helper Phage, Cytiva
Nucleic Acid Storage Systems
M13 phage for the GST expression system.
  • For phage rescue
  • Colorless to light yellow liquid
  • Concentration >5×10¹⁰ pfu/ml
The M13KO7 helper phage is an M13mp1 derivative containing a modified gene 11 (G changed to T at position 6,125, giving a met to ile change at codon 40 of the gene 11 protein), an insertion of the origin of replication of p15A and the kanamycin resistance gene from Tn903 at the Ava I site (position 5,825).