Anti-IgG Donkey Antibody (Biotin)

Supplier: Cytiva
RPN1004
95017-418EA 459.88 USD
95017-418
Anti-IgG Donkey Antibody (Biotin)
Antibodies
The antibodies are supplied with dilution guidelines for relevant applications.

  • Soluble in cold water and hot water

The antibodies are supplied with dilution guidelines for relevant applications and are stored at 2°C to 8°C.

Type: Secondary
Antigen: Rabbit Ig
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Donkey
Isotype: none
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR