Buffer Reservoir, Cytiva
Supplier: Cytiva
18-3216-03
95017-305PK 73.66 USD
95017-305
Buffer Reservoir, Cytiva
Reagent Reservoirs
Fast, affordable, high-recovery desalting and buffer exchange, as well as removal of low-molecular weight compounds packed with Sephadex G-25 Medium, LabMate buffer reservoir makes it easier and more convenient to equilibrate PD-10 columns.
  • Easy to use and equilibrate, apply sample, elute
  • Buffer reservoir for easy equilibration
  • No wetted part
Packaging: Pack size 10