Sodium dodecyl sulfate (SDS), PlusOne™

Supplier: Cytiva
Danger

17-1313-01
95017-116EA 83.92 USD
95017-116
Sodium dodecyl sulfate (SDS), PlusOne™
Sodium dodecyl sulphate (SDS)

SDS is an ionic detergent used to solubilize proteins. The PlusOne SDS reagent is carefully controlled for carbon chain length and overall quality.


  • pH Range: 6.5 - 8.5
  • Partially soluble in cold water, hot water and methanol,
  • Very slightly soluble in acetone, insoluble in diethyl ether

Formula: CH₃(CH₂)₁₁OSO₃Na
Melting Pt: 204…207 °C
Storage Temperature: Ambient
MDL Number: MFCD00036175
CAS Number: 151-21-3
UN: 8027
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR