illustra™ Sephadex® G-25 Grade DNA Clean Up, Cytiva
Supplier: Cytiva
17-0572-02
95016-892EA 647.66 USD
95016-892
illustra™ Sephadex® G-25 Grade DNA Clean Up, Cytiva
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
For purification of DNA (10 bases in length) from small molecules by gel filtration.
  • Tested to ensure reproducibly high recovery of DNA
  • Used in MicroSpin G-25 Columns and NAP Columns
  • Enables self preparation of columns for nucleic acid purification