Sulfuric acid solution 10% (w/w) FCC

Supplier: Columbus Chemical Industries, Inc
Warning

986900-22501 986900-21001
95000-002EA 1950 USD
95000-002 95000-000
Sulfuric acid solution 10% (w/w) FCC
Sulfuric acid
Formula: H₂SO₄
MW: 98.08
Boiling Pt: 330
Melting Pt: 10.38
Density: 1.066 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064589
CAS Number: 7664-93-9
EINECS: 231-639-5
UN: 2796
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR