Three Star™ Flexible Tubing/Cable Cutter, Excelta
Supplier: Excelta
51-T
94024-286EA 42.31 USD
94024-286
Three Star™ Flexible Tubing/Cable Cutter, Excelta
Cutters
This stainless-steel cutter provides a smooth clean cut on plastic tubing and soft cable.
  • This tool features with ergonomic plastic molded handle
The locking feature helps in keeping the jaws closed while not in use.