SATWipes® SWNW0013 Series Presaturated Wipes, Contec®
Supplier: Contec
SWNW0013
94002-780CS 204.91 USD
94002-780
SATWipes® SWNW0013 Series Presaturated Wipes, Contec®
Wipes Cleanroom Wipes Presaturated Cleanroom Wipes
SATWipes® SWNW0013 series wipes consist of a roll of (700) 7×9" saturated wipes which are dispensed through a center-pull container.
  • Roll of wipes are conviently dispensed through a canister
  • Presaturated for quick clean-ups
  • Strong fabric is also low linting
The wipes are made of 50% polyester and 50% rayon nonwoven material, creating a soft, durable, and clean wipe. The fabric is strong, sorbent, and low linting. The saturation of 70% alcohol and 30% deionized water provides a safe and mild cleaning solution.