Autoclave Cleaning Brush Kit, Sklar
Supplier: Sklar
10-1424
94002-700EA 441.18 USD
94002-700
Autoclave Cleaning Brush Kit, Sklar
Brushes
The Autoclave Cleaning Brush Kit contains one telescopic handle which extends from 61–122cm (24–48"), one large 13.9x19cm (5.5x7.5") cleaning brush with round corner bristles, and one hand brush with a 11.4cm (4.5") handle.
All components are reusable, nonsterile, and latex-free.