Autoclave Cleaning Brush, Sklar
Supplier: Sklar
10-1422
94002-698PK 280.68 USD
94002-698
Autoclave Cleaning Brush, Sklar
Brushes
The Autoclave Cleaning Brush is designed for use on autoclave chambers and doors, sinks, and carts.
  • Nonsterile
  • Latex-free
The brush head measures 7.6x8.9cm (3x3.25"), and features white medical grade nylon bristles. Overall length is 48.9cm (19.25").