BD BBL™ Isolator Pack Plated Media, BD Diagnostics
Supplier: BD
BD BBL™ prepared agar plates in isolator packs are plated media in gamma-irradiated packs that are specially designed for isolator systems
- Specially designed for isolator systems
- Validated for performance and sterility according to the Association for the Advancement of Medical Instrumentation (AAMI) guidelines
To use, place wrapped package of plates in the isolator before running the vaporized hydrogen peroxide decontamination cycle. When the isolator aeration cycle has finished, remove the two outermost wrappings, then the sterile inner wrapping. Use the included inner bag to transport plates from your critical environment. Storage temperature: 2 to 8 °C.
The multi-wrap packaging protects media from vaporized hydrogen peroxide exposure.
Ordering information: Each pack or case contains a signed, lot-specific Certificate of Analysis.