Difco™ m FC Broth Base, BD Diagnostics
Supplier: BD
288330
90004-086EA 144.42 USD
90004-086 90004-088
Difco™ m FC Broth Base, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Use with Rosolic Acid For detection and enumeration of fecal coliforms by the membrane filter technique at elevated temperatures.