Difco™ D-Nase Test Agar, BD
Supplier: BD
263220
90000-108EA 264.62 USD
90000-108 90003-736
Difco™ D-Nase Test Agar, BD
Media for Microbiology Prepared Media for Microbiology
For differentiating microorganisms based on deoxyribonuclease activity.