Difco™ DRBC Agar, BD
Supplier: BD
258710
90003-540EA 234.42 USD
90003-540
Difco™ DRBC Agar, BD
Media for Microbiology Dehydrated Media for Microbiology
Difco™ DRBC Agar (Dichloran Rose Bengal Chloramphenicol) supports good growth of yeasts and molds while inhibiting bacteria and the spreading of rapidly growing molds.