Difco™ 2XYT Medium, BD Diagnostics
Supplier: BD
244020
90003-330EA 163.95 USD
90003-330 90003-328
Difco™ 2XYT Medium, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
For growth and propagation of E
  • Dehydrated culture media
  • Sequencing and phage display research
  • Provides nitrogen and growth factor
A nutritionally rich growth medium designed for growth of recombinant strains of Escherichia coli. Also used for propagation of M13 bacteriophage for sequencing and phage display research. The components of Difco 2xYT Medium provide nitrogen and growth factors that allow bacteriophage to reproduce in large quantities without exhausting the host.
coli infected with single strand filamentous bacteriophage M13.