BD Taxo™ Novobiocin Discs, BD Diagnostic Systems
Supplier: BD
231750
90002-578PK 110.53 USD
90002-578
BD Taxo™ Novobiocin Discs, BD Diagnostic Systems
Microbiology Test Systems Microorganism Tests
Taxo™ Differentiation Discs Novobiocin are recommended for the differentiation of coagulase negative staphylococci (e.g., Staphylococcus saprophyticus) based on novobiocin resistance.