BBL™ DrySlide™ Nitrocefin, BD
Supplier: BD
231749
90002-576PK 556.41 USD
90002-576
BBL™ DrySlide™ Nitrocefin, BD
Slides Chamber Slides
A disposable slide containing a reagent cassette impregnated with nitrocefin for the rapid qualitative detection of beta-lactamase producing bacteria.
  • Test results in 60 minutes or less