DrySlide™ Indole Slide, BD Biosciences
Supplier: BD
231748
90002-574PK 1001.72 USD
90002-574
DrySlide™ Indole Slide, BD Biosciences
Slides Chamber Slides
Disposable slide employs a dry reagent cassette impregnated with 5% p-dimethylaminobensaldehyde (DMABA) for the rapid spot-identification of indole producing bacteria from primary cultures
Divided into four large segments to enable multiple tests on one slide. Provides results in just 30 seconds