BD BBL™ Taxo™ Differentiation Discs for Haemophilus Species, BD Diagnostic
Supplier: BD
231727
90002-550PK 119.21 USD
90002-550
BD BBL™ Taxo™ Differentiation Discs for Haemophilus Species, BD Diagnostic
Microbiology Test Systems Microorganism Tests
BD BBL™ Taxo™ Differentiation Discs ideal for differentiating Haemophilus SPP.
  • Ideal for differentiating Haemophilus SPP
  • Based on X or V factors or both
Differentiation is based on their requirements for X or V factors or both.