BBL™ Cefinase Paper Disc,¼, Impregnated with Nitrocefin, BD
Supplier: BD
231650
90002-462PK 277.02 USD
90002-462
BBL™ Cefinase Paper Disc,¼, Impregnated with Nitrocefin, BD
Test Paper Indicator Paper
Paper Discs for the Detection of Beta-Lactamase Enzymes
Packaging is Latex free.