BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, BD Diagnostics
Supplier: BD
230918 230942 231034 231227 231267 231299 231328 231342 231344 231353 231536 231539 231541 231574 231601 231632 231645 231672 231678 231691 231696 231704 231706 231762 230886 230906 231030 231149 231321 231352 231569 231573 231605 231674 231693 231703 231754 232197 230874 230998 231250 231292 231301 231311 231313 231314 231315 231319 231324 231568 231608 231609 231619 231666 231705 231293 231544 231603 231644 231673 231692 231694 232174 232219 231251 231621 232175 231634 231640 231647 231648 231649 231717 291255 231634 231640 231647 231648 231649 232231 232218 231755 231758 232208
90002-100EA 48.22 USD
90002-100 90002-104 90002-112 90002-216 90002-232 90002-262 90002-290 90002-296 90002-300 90002-310 90002-324 90002-326 90002-328 90002-358 90002-390 90002-430 90002-452 90002-492 90002-500 90002-512 90002-522 90002-530 90002-534 90002-544 90002-582 90002-090 90002-094 90002-110 90002-210 90002-284 90002-308 90002-348 90002-356 90002-394 90002-473 90002-497 90002-516 90002-528 95027-260 96062-002 90002-086 90002-106 90002-224 90002-252 90002-264 90002-270 90002-274 90002-276 90002-278 90002-280 90002-286 90002-346 90002-400 90002-403 90002-416 90002-488 90002-532 95027-258 90002-254 90002-330 90002-392 90002-450 90002-468 90002-494 90002-515 90002-518 90002-596 95053-328 90002-226 90002-420 90002-598 90002-434 90002-444 90002-456 90002-458 90002-460 90002-542 90004-144 96062-006 BB4331634 BB4331640 BB4331647 BB4331648 BB4331649 BD232231 75877-344 76070-162 76070-164 76070-166
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, BD Diagnostics
Microbiology Test Systems Microorganism Tests
The 6 mm diameter (¹/₄") discs are made from high quality absorbent paper that has been impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents. Discs are clearly marked on both sides with letters and numbers designating the agent and the drug content.
Ordering information: Discs are packaged 50 discs per cartridge. The last disc in each cartridge is marked with an 'X' and contains the agent as coded. Cartridges are for use in BD BBL™ Sensi-Disc™ dispensers.